Läs mer
Sodium Tricyanomethanide, TRC
Produktkod. 30135853
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135853 500 mg
30138044 50 mg
30006461 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135853 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS673658500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36603-80-2
Molekylformel C4 N3 . Na
Molekylvikt (g/mol) 113.053
IUPAC-namn sodium;methanetricarbonitrile
LEDER [Na+].N#C[C-](C#N)C#N
Specifikationer
Kemiskt namn eller material Sodium Tricyanomethanide
Kvantitet 500 mg
InChI formel InChI=1S/C4N3.Na/c5-1-4(2-6)3-7;/q-1;+1
Formel vikt 112.999