missing translation for 'onlineSavingsMsg'
Läs mer

Sodium Trifluoroacetate-13C, TRC

Produktkod. 30414777
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30414777 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30414777 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCS55446010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Sodium Trifluoroacetate-1-13C
CAS 286425-32-9
Molekylformel 13C C F3 O2 . Na
Kvantitet 10 mg
Synonym Acetic-1-13C acid','2,2,2-trifluoro-','sodium salt (9CI), 2,2,2-Trifluoroacetic-1-13C acid sodium salt, Trifluoroacetic-1-13C acid sodium salt, Sodium 2,2,2-trifluoroacetate-1-13C, Sodium trifluoroacetate-1-13C
InChI formel InChI=1S/C2HF3O2.Na/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1/i1+1;
LEDER [Na+].[O-][13C](=O)C(F)(F)F
IUPAC-namn sodium;2,2,2-trifluoroacetate
Molekylvikt (g/mol) 136.998
Formel vikt 136.978
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.