missing translation for 'onlineSavingsMsg'
Learn More

Sodium trifluoromethanesulfinate, 98%, Thermo Scientific Chemicals

Catalog Number p-7077526
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
2926-29-6
CF3NaO2S
156.06
KAVUKAXLXGRUCD-UHFFFAOYSA-M
sodium trifluoromethanesulfinate, sodium trifluoromethanesulphinate, sodium trifluoro-methanesulfinate, trifluoromethanesulfinic acid sodium salt, methanesulfinic acid, trifluoro-, sodium salt, langlois reagent, chf3o2s.na, acmc-209h7v, ksc563a2r, sodium trifluoromethylsulfinate
23690734
sodium;trifluoromethanesulfinate
C(F)(F)(F)S(=O)[O-].[Na+]
This item is not returnable. View return policy
2926-29-6
CF3NaO2S
156.06
KAVUKAXLXGRUCD-UHFFFAOYSA-M
sodium trifluoromethanesulfinate, sodium trifluoromethanesulphinate, sodium trifluoro-methanesulfinate, trifluoromethanesulfinic acid sodium salt, methanesulfinic acid, trifluoro-, sodium salt, langlois reagent, chf3o2s.na, acmc-209h7v, ksc563a2r, sodium trifluoromethylsulfinate
23690734
sodium;trifluoromethanesulfinate
C(F)(F)(F)S(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 350.0°C
Quantity 1 g
Assay Percent Range 98%
Linear Formula CF3SO2Na
Solubility Information Solubility in water: soluble.
Formula Weight 156.06
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Sodium trifluoromethanesulfinate
compliance-icons
Product Identifier
  • Sodium trifluoromethanesulfinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'