Learn More
Sofosbuvir impurity K, MedChemExpress
Product Code. 16421248
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421248 5 mg 5mg
16402469 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16421248 Supplier Medchem Express Supplier No. HYI0515
 Restricted Item
Beräknad leverans: 26-11-2026
MedChemExpress Sofosbuvir impurity K, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Sofosbuvir impurity K, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Specifications
Chemical Name or Material Sofosbuvir impurity K
CAS 1496552-51-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H29ClN3O9P
Quantity 5 mg
SMILES O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](O)[C@](Cl)(C)[C@H](N2C(NC(C=C2)=O)=O)O1)OC3=CC=CC=C3
Molecular Weight (g/mol) 545.91
Formula Weight 545.91
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus