Learn More
Megro™ SOFT-JECT™ Disposable Syringes
A 3-part series of syringe models with additional black rubber plugs for more comfortable handling and better reading accuracy.
Brand: Megro™ 5100.000V0
Additional Details : Weight : 0.50000kg
Description
- Larger diameters and higher sealing lips guarantees a better density of the syringe
- Second brake ring plate at the piston rod provides better and exact sliding of the piston rod
- Latex-free
- Clear for excellent optical clarity and visibility of contents
- Piston and cylinder of polypropylene, piston plug of synthetic polyisoprene-rubber
- Luer attachment
Specifications
Polypropylene (Barrel), Polypropylene (Plunger) | |
Comfortable handling, better accuracy | |
Yes | |
10 mL | |
Yes |
Transparent | |
Yes | |
100/Pk. | |
Luer |