Läs mer
Solabegron Hydrochloride, TRC
Produktkod. 30416762
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30416762 1 mg
1 options
Produktkod. 30416762 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6765101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Solabegron Hydrochloride
CAS 451470-34-1
Molekylformel C23H23ClN2O3.HCl
Kvantitet 1 mg
Synonym 3'-[[2-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]ethyl]amino]-[1,1'-biphenyl]-3-carboxylic Acid Hydrochloride, 3'-[[2-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]ethyl]amino]-[1,1'-biphenyl]-3-carboxylic Acid Monohydrochloride, (R)-3'-((2-((2-(3-Chlorophenyl)-2-hydroxyethyl)amino)ethyl)amino)-[1,1'-biphenyl]-3-carboxylic Acid Hydrochloride, GW 427353B
InChI formel InChI=1S/C23H23ClN2O3.ClH/c24-20-8-2-6-18(13-20)22(27)15-25-10-11-26-21-9-3-5-17(14-21)16-4-1-7-19(12-16)23(28)29;/h1-9,12-14,22,25-27H,10-11,15H2,(H,28,29);1H/t22-;/m0./s1
LEDER Cl.O[C@@H](CNCCNc1cccc(c1)c2cccc(c2)C(=O)O)c3cccc(Cl)c3
IUPAC-namn 3-[3-[2-[[(2R)-2-(3-chlorophenyl)-2-hydroxyethyl]amino]ethylamino]phenyl]benzoic acid;hydrochloride
Molekylvikt (g/mol) 447.4
Formel vikt 446.1164