Läs mer
Solanesyl Bromide, TRC
Produktkod. 30410406
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410406 25 mg
30410078 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30410406 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS67652025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52610-77-2
Molekylformel C45H73Br
Molekylvikt (g/mol) 693.97
Synonym trans-Solanesyl Bromide, (2E,6E,10E,14E,18E,22E,26E,30E)-1-Bromo-3,7,11,15,19,23,27,31,35-nonamethyl-, 2,6,10,14,18,22,26,30,34-hexatriacontanonaene
IUPAC-namn (2E,6E,10E,14E,18E,22E,26E,30E)-1-bromo-3,7,11,15,19,23,27,31,35-nonamethylhexatriaconta-2,6,10,14,18,22,26,30,34-nonaene
LEDER CC(=CCC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CBr)\C)\C)\C)\C)\C)\C)\C)\C)C
Specifikationer
Kemiskt namn eller material Solanesyl Bromide
Kvantitet 25 mg
InChI formel InChI=1S/C45H73Br/c1-37(2)19-11-20-38(3)21-12-22-39(4)23-13-24-40(5)25-14-26-41(6)27-15-28-42(7)29-16-30-43(8)31-17-32-44(9)33-18-34-45(10)35-36-46/h19,21,23,25,27,29,31,33,35H,11-18,20,22,24,26,28,30,32,34,36H2,1-10H3/b38-21+,39-23+,40-25+,41-27+,42-29+,43-31+,44-33+,45-35+
Formel vikt 692.4896