Läs mer
Solasodine Hydrochloride, TRC
Produktkod. 30413330
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30413330 10 mg
1 options
Produktkod. 30413330 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS67655510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Solasodine Hydrochloride
CAS 6106-33-8
Molekylformel C27H43NO2 . HCl
Kvantitet 10 mg
Synonym Solasod-5-en-3β-ol Hydrochloride (8CI), (3β,22α,25R)- (9CI)-Spirosol-5-en-3-ol Hydrochloride, NSC 35543
InChI formel InChI=1S/C27H43NO2.ClH/c1-16-7-12-27(28-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(29)8-10-25(18,3)21(20)9-11-26(22,24)4;/h5,16-17,19-24,28-29H,6-15H2,1-4H3;1H/t16-,17+,19+,20-,21+,22+,23+,24+,25+,26+,27-;/m1./s1
LEDER Cl.C[C@@H]1CC[C@@]2(NC1)O[C@H]3C[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC[C@]4(C)[C@H]3[C@@H]2C
Molekylvikt (g/mol) 413.64
Formel vikt 449.3061