Läs mer
Solasodine, MedChemExpress
Produktkod. 16498966
Encompass_Preferred
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
16498966 100 mg
1 options
Produktkod. 16498966 Leverantör Medchem Express Leverantörsnummer HYN0068
MedChemExpress Solasodine (Purapuridine) is a steroidal alkaloid that occurs in plants of the Solanaceae family. Solasodine has neuroprotective, antifungal, hypotensive, anticancer, antiatherosclerotic, antiandrogenic and anti-inflammatory activities.
Solasodine (Purapuridine) is a steroidal alkaloid that occurs in plants of the Solanaceae family. Solasodine has neuroprotective, antifungal, hypotensive, anticancer, antiatherosclerotic, antiandrogenic and anti-inflammatory activities.
Specifikationer
Kemiskt namn eller material Solasodine
Fysisk form Powder
CAS 126-17-0
Färg Off-White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C27H43NO2
För användning med (applikation) Cancer-programmed cell death
Synonym Purapuridine Solancarpidine Solasodin
Löslighetsinformation DMSO : 2.6 mg/mL (6.29 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.62 mM; Need ultrasonic)
LEDER C[C@H](CC1)CN[C@]21O[C@@]3([H])C[C@@]4([H])[C@]5([H])CC=C6C[C@@H](O)CC[C@]6(C)[C@@]5([H])CC[C@]4(C)[C@@]3([H])[C@@H]2C
Molekylvikt (g/mol) 413.64
Formel vikt 413.64
Procent renhet 95.0%