Läs mer
Solerole, TRC
Product Code. 30411653
Change view
Click to view available options
Kvantitet:
250 mg
Product Code. Kvantitet
30411653 250 mg
1 options
Product Code. 30411653 Supplier Toronto Research Chemicals Supplier No. TRCS676630250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Solerole
CAS 27610-27-1
Molekylformel C6H10O3
Kvantitet 250 mg
Synonym 4,5-Dihydroxyhexanoic Acid gamma-Lactone, Dihydro-5-(1-hydroxyethyl)-2(3H)-furanone
InChI formel InChI=1S/C6H10O3/c1-4(7)5-2-3-6(8)9-5/h4-5,7H,2-3H2,1H3
LEDER CC(O)C1CCC(=O)O1
IUPAC-namn 5-(1-hydroxyethyl)oxolan-2-one
Molekylvikt (g/mol) 130.14
Formel vikt 130.063