Learn More
Brady™ Solid Brass Housing and Bracket Padlock
Brand:  Brady™ 805830
Additional Details : Weight : 1.40700kg
Product Code. 11731376
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Brass Padlock
Brass
7.0 mm
73 mm
Stainless Steel
6 Pk.
49 mm
Chrome
Brass Padlocks
49.00 x 73.00 mm
27.0 mm
5
49 mm