Läs mer
Solifenacin Hydrochloride, TRC
Produktkod. 30415957
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30415957 10 mg
1 options
Produktkod. 30415957 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS67670010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Solifenacin Hydrochloride
CAS 180468-39-7
Molekylformel C23 H26 N2 O2 . Cl H
Kvantitet 10 mg
Synonym 2(1H)-Isoquinolinecarboxylic acid','3,4-dihydro-1-phenyl-','(3R)-1-azabicyclo[2.2.2]oct-3-yl ester','hydrochloride (1:1)','(1S)-, 2(1H)-Isoquinolinecarboxylic acid','3,4-dihydro-1-phenyl-','(3R)-1-azabicyclo[2.2.2]oct-3-yl ester','monohydrochloride','(1S)- (9CI), 2(1H)-Isoquinolinecarboxylic acid','3,4-dihydro-1-phenyl-','1-azabicyclo[2.2.2]oct-3-yl ester','monohydrochloride','[R-(R*,S*)]-, Solifenacin hydrochloride
InChI formel InChI=1S/C23H26N2O2.ClH/c26-23(27-21-16-24-13-10-18(21)11-14-24)25-15-12-17-6-4-5-9-20(17)22(25)19-7-2-1-3-8-19;/h1-9,18,21-22H,10-16H2;1H/t21-,22-;/m0./s1
LEDER Cl.O=C(O[C@H]1CN2CCC1CC2)N3CCc4ccccc4[C@@H]3c5ccccc5
IUPAC-namn [(3R)-1-azabicyclo[2.2.2]octan-3-yl] (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate;hydrochloride
Molekylvikt (g/mol) 398.93
Formel vikt 398.1761