missing translation for 'onlineSavingsMsg'
Läs mer

Solifenacin N-Oxide, Mikromol

Produktkod. 30458229
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30458229 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30458229 Leverantör Mikromol Leverantörsnummer MM1342.040025

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Solifenacin N-Oxide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Solifenacin N-Oxide
CAS 180272-28-0
Analyt- eller komponentnamn Solifenacin N-Oxide
Frakt skick Ice pack (-20°C)
Molekylformel C23 H26 N2 O3
Certifikat och efterlevnad ISO 9001
Synonym 2(1H)-Isoquinolinecarboxylic acid, 3,4-dihydro-1-phenyl-, (3R)-1-oxido-1-azabicyclo[2.2.2]oct-3-yl ester, (1S)-, Solifenacin N1-oxide, Solifenacin Succinate Impurity I, Solifenacin Impurity I
InChI formel InChI=1S/C23H26N2O3/c26-23(28-21-16-25(27)14-11-18(21)12-15-25)24-13-10-17-6-4-5-9-20(17)22(24)19-7-2-1-3-8-19/h1-9,18,21-22H,10-16H2/t18?,21-,22-,25?/m0/s1
Lösningstyp Neat
LEDER [O-][N+]12CCC(CC1)[C@H](C2)OC(=O)N3CCc4ccccc4[C@@H]3c5ccccc5
IUPAC-namn [(3R)-1-oxido-1-azoniabicyclo[2.2.2]octan-3-yl] (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate
Molekylvikt (g/mol) 378.46
Kvantitet 25 mg
Formel vikt 378.1943 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.