missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Ansell Edmont™ Solvex™ 37-185 Series Nitrile Chemical Resistant Gloves
Handla allt Ansell Produkter
Klicka för att se tillgängliga alternativ
Storlek:
8
9
10
11
Förpackningsstorlek:
Paket med 2
Beskrivning
- Chemical and liquid protection gloves
- High performance nitrile compound provides better combination of chemical resistance and strength
- Offers superior snag, puncture and abrasion protection over rubber or neoprene gloves
- Fully reusable, with an unequaled abrasion resistance, it provides better comfort for the wearer
- Ideal choice for safe handling in wide range of work environments where harsh chemicals are present
- FDA Food Approved
- Suitable for heavy-duty cleaning applications
- High levels of flexibility, comfort and dexterity
- No inner cotton flocking, reducing risk of lint contamination and making it ideal for production areas sensitive to the introduction of external impurities
- Sand patch finish gives the glove smooth surface that decreases indirect costs thanks to fewer rejects of fragile parts
Applications:
- All food processing applications that have food contact
- Filling, blending and charging of raw materials
- Forging operations
- General pumping work
- Handling incoming goods
- Handling outgoing goods: cans, vessels, bulk pack and cartons
- Loading and unloading process equipment
- Opening and draining pumps valves or lines
- Opening furnaces draining pumps valves or lines and crackers BTX process
- Painting
- Supervision of running operation
- Testing
- Transferring liquids and solids between vessels and tanks and process equipment
Specifikationer
Specifikationer
| Certifikat och efterlevnad | EN 420:2003 + A1:2009 and EN 388:2003,EN 374:2007 |
| Färg | Green |
| Latexfri | Yes |
| Material | Nitrile |
| Produkttyp | Immersion Gloves |
| Storlek | 9 |
| Stil | Grip design: Sandpatch |
| Tjocklek (metrisk) | 0.56 mm |
| För användning med (applikation) | Food |
| Handske manschettstil | Gauntlet |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering