Läs mer
Sombrevin, TRC
Produktkod. 17961765
Change view
Kvantitet:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17961765 25 mg
17922755 1 g
17953045 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17961765 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS67676525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1421-14-3
Molekylformel C18 H27 N O5
Molekylvikt (g/mol) 337.41
Synonym Sombrevin, 4-[2-(Diethylamino)-2-oxoethoxy]-3-methoxybenzeneacetic Acid Propyl Ester, 13245RP, 2180TH, BAY 1420, Epontol, FBA 1420, Fabantol, Fabontal, Propanidide, Propantan, Propyl 4-(N,N-Diethylcarbamidomethoxy)-3-methoxy-phenylacetate, Propyl [4-[(Diethylcarbamoyl)methoxy]-3-methoxyphenyl]acetate, TH 2180, WH 5668, [4-[(Diethylcarbamoyl)methoxy]-3-methoxyphenyl]acetic Acid Propyl Ester
IUPAC-namn propyl 2-[4-[2-(diethylamino)-2-oxo-ethoxy]-3-methoxy-phenyl]acetate
LEDER CCCOC(=O)Cc1ccc(OCC(=O)N(CC)CC)c(OC)c1
Specifikationer
Kemiskt namn eller material Propanidid
Kvantitet 25 mg
InChI formel InChI=1S/C18H27NO5/c1-5-10-23-18(21)12-14-8-9-15(16(11-14)22-4)24-13-17(20)19(6-2)7-3/h8-9,11H,5-7,10,12-13H2,1-4H3
Formel vikt 337.1889