missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ SON Polyclonal Antibody

Produktkod. 15762741 Handla allt Thermo Scientific Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15762741 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15762741 Leverantör Invitrogen™ Leverantörsnummer PA554814

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: TEVAIESTPM ILESSIMSSH VMKGINLSSG DQNLAPEIGM QEIALHSGEE PHAEEHLKGD FYESEHGINI DLNINNHLIA KEMEHNTVCA AGTSPVGEIG EEKILPTSET KQRTVLDTYP GVSEADAGET LSSTGP Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 60%.

The protein encoded by this gene binds to a specific DNA sequence upstream of the upstream regulatory sequence of the core promoter and second enhancer of human hepatitis B virus (HBV). Through this binding, it represses HBV core promoter activity, transcription of HBV genes, and production of HBV virions. The protein shows sequence similarities with other DNA-binding structural proteins such as gallin, oncoproteins of the MYC family, and the oncoprotein MOS. It may also be involved in protecting cells from apoptosis and in pre-mRNA splicing. Several transcript variants encoding different isoforms have been described for this gene, but the full-length nature of only two of them has been determined. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen SON
Användningsområden Immunohistochemistry (Paraffin), Immunocytochemistry
Klassificering Polyclonal
Koncentration 0.10 mg/mL
Konjugera Unconjugated
Formulering PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gen SON
Genaccessionsnr. P18583
Gene Alias 2900011L12Rik; AA409051; AU067731; BASS1; Bax antagonist selected in Saccharomyces 1; C21orf50; C81487; DBP5; DBP-5; HSPC310; HSPC312; KIAA1019; mKIAA1019; negative regulatory element-binding protein; NRE-binding protein; NREBP; Protein DBP-5; Protein SON; SON; Son cell proliferation protein; Son DNA binding protein; SON DNA-binding protein; SON3
Gensymboler SON
Värd art Rabbit
Immunogen Recombinant protein corresponding to Human SON. Recombinant protein control fragment (Product #RP-91770).
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 6651
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.