Läs mer
Sonidegib, MedChemExpress
Produktkod. 16487126
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16487126 100 mg
16483127 50 mg
16457088 10 mg
16409637 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16487126 Leverantör Medchem Express Leverantörsnummer HY16582A
MedChemExpress Sonidegib (Erismodegib) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively.
Sonidegib (Erismodegib) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively.
Specifikationer
Kemiskt namn eller material Sonidegib
CAS 956697-53-3
Färg Purple
Anteckningar om renhetsgrad Research
Molekylformel C26H26F3N3O3
Kvantitet 100 mg
Synonym Erismodegib LDE225 NVP-LDE225
Löslighetsinformation DMSO : 50 mg/mL (102.99 mM; Need ultrasonic)
LEDER O=C(C1=C(C)C(C(C=C2)=CC=C2OC(F)(F)F)=CC=C1)NC3=CC=C(N=C3)N4C[C@@H](C)O[C@@H](C)C4
Molekylvikt (g/mol) 485.5
Formel vikt 485.5
Procent renhet 99.64%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death