Läs mer
Sorafenib Tosylate, TRC
Produktkod. 30136661
Change view
Kvantitet:
100 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136661 10 g
30003256 1 g
30008452 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136661 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS67684510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 475207-59-1
Molekylformel C21 H16 Cl F3 N4 O3 . C7 H8 O3 S
Molekylvikt (g/mol) 637.03
Synonym 4-[4-[[[[4-chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide 4-methylbenzenesulfonate (1:1), 2-Pyridinecarboxamide, 4-[4-[[[[4-chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-, mono(4-methylbenzenesulfonate) (9CI), BAY 43-9006 mono-p-tosylate, BAY 54-9085, Nexavar, Sorafenib p-toluenesulfonate, Sorafenib tosylate
IUPAC-namn 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]-N-methylpyridine-2-carboxamide;4-methylbenzenesulfonic acid
LEDER CNC(=O)c1cc(Oc2ccc(NC(=O)Nc3ccc(Cl)c(c3)C(F)(F)F)cc2)ccn1.Cc4ccc(cc4)S(=O)(=O)O
Specifikationer
Kemiskt namn eller material Sorafenib Tosylate
Kvantitet 10 g
InChI formel InChI=1S/C21H16ClF3N4O3.C7H8O3S/c1-26-19(30)18-11-15(8-9-27-18)32-14-5-2-12(3-6-14)28-20(31)29-13-4-7-17(22)16(10-13)21(23,24)25;1-6-2-4-7(5-3-6)11(8,9)10/h2-11H,1H3,(H,26,30)(H2,28,29,31);2-5H,1H3,(H,8,9,10)
Formel vikt 636.1057