Läs mer
Sorbitan Monooleate (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Sorbitan Monooleate (Technical Grade) |
| CAS | 1338-43-8 |
| Molekylformel | C24H44O6 |
| Kvantitet | 250 mL |
| Synonym | (Z)-Mono-9-octadecenoate Sorbitan, Alkamuls SMO, Arlacel 80, Armotan MO, Atlas G 946, Atmer 105, Atmer 105V, Crill 4, Dehymuls SMO, Disponil 100, E 494, Emasol 410, Emasol O 10, Emasol O 10F, Emasol O 10V, Emsorb 2500, G 946, Glycomul O, Ionet S 80, Kemmat S 80, Kosteran O 1, Kosteran O 1VL, Liposorb 80, Lonzest SMO, MO 33F, Monemul 80, Monodehydrosorbitol Monooleate, Monopol SP 1, Montane 80, Montane 80 VGA, Montane 80VGPha, Newcol 80, Nikkol SO 10, Nikkol SO 10R, Nikkol SO 10T, Nikkol SO 10V, Nissan Nonion OP 80, Nissan Nonion OP 80R, Nofable SO 851S, Nonion 80R, Nonion OP 80, Nonion OP 80R, O 250, OP 80, OP 80R, PAS 80, Pionin D 935, Poem O 80V, Protachem SMO, Rheodol AO 10, Rheodol AO 10V, Rheodol SP-O 10, Rheodol SP-O 10V, Rikemal O 250, S 270, S 271, S 80, S 84, S-MAX 80, S-Maz 80K, SM 80, SO 10V, SO 851S, SP 010, SP 80, SP-O 10, SP-O 10V, Sorbester P 17, Sorbitan O, Sorbitan Monooleic Acid Ester, Sorbon S 80, Sorgen 40, Sorgen 40A, Sorgen 40V, Span 280, Span 80, Span S 80, Sunsoft 81S, Super 80, Sylvan S 80 |
| InChI formel | InChI=1S/C24H44O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(27)29-19-21(26)24-23(28)20(25)18-30-24/h9-10,20-21,23-26,28H,2-8,11-19H2,1H3/b10-9-/t20-,21+,23+,24+/m0/s1 |
| LEDER | CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@@H](O)[C@H]1OC[C@H](O)[C@H]1O |
| IUPAC-namn | [(2R)-2-[(2R,3R,4S)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl] (Z)-octadec-9-enoate |
| Molekylvikt (g/mol) | 428.6 |
| Formel vikt | 428.3138 |