Learn More
SORBS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555656
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DSSPLLNEVS SSLIGTDSQA FPSVSKPSSA YPSTTIVNPT IVLLQHNREQ QKRLSSLSDP VSERRVGEQD SAPTQEKPTS PGKAIEKRAK DDSRRVVKST QDLSDVSMDE VGIPLRNTER SKDWYKTMFK QIHKLNR Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
SORBS1 Polyclonal antibody specifically detects SORBS1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SORBS1 | |
Polyclonal | |
Unconjugated | |
SORBS1 | |
2310065E01Rik; 9530001P15Rik; CAP; c-Cbl associated protein; c-Cbl-associated protein; DKFZp451C066; DKFZP586P1422; Fas-ligand associated factor 2; FLAF2; FLJ12406; I79_015895; KIAA0894; KIAA1296; mKIAA1296; OTTHUMP00000020142; OTTHUMP00000020143; OTTHUMP00000020144; OTTHUMP00000020145; Ponsin; R85FL; SH3 domain protein 5; Sh3d5; SH3P12; SORB1; sorbin and SH3 domain containing 1; sorbin and SH3 domain-containing protein 1; Sorbs1; sorbs1 {ECO:0000250; UniProtKB:Q62417} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10580 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BX66 | |
SORBS1 | |
Recombinant protein corresponding to Human SORBS1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only