missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23925-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may play a role in tumorigenesis. A similar protein in mice is involved in the regulation of the wingless-type MMTV integration site family (Wnt) pathway.Specifications
SOX7 | |
Polyclonal | |
Unconjugated | |
SOX7 | |
SOX7, Transcription factor SOX 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20680, 83595 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P40646, Q9BT81 | |
SOX7 | |
SOX7 Fusion Protein Ag10323 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |