Learn More
3M™ Soy Protein Rapid Kit L25SOY
Intended for screening for the presence of soy proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Brand: 3M™ 7100151326
4548.13 SEK valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Additional Details : Weight : 0.30200kg
Description
- Results visible at 11 ± 1 minute after application of the sample.
- Rapid, qualitative immunochromatographic lateral flow test cartridges designed for the detection of allergenic food proteins.
- Designed to detect both processed and unprocessed allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
Specifications
Microbiology Media | |
Allergen | |
25/Cs. |
Safety and Handling
- 3M Soy Protein Rapid Kit
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : ethanol, glycerol, urea, sodium chloride