Läs mer
Soybean Oil Epoxide, TRC
Produktkod. 30409147
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30409147 10 g
1 options
Produktkod. 30409147 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS67786310G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Soybean oil','epoxidized
CAS 8013-07-8
Molekylformel C57H98O12
Kvantitet 10 g
Synonym ESBO','Soybean oil','epoxidized','Epoxidized soya bean oil
InChI formel InChI=1S/C57H98O12/c1-4-7-19-28-43-49(64-43)37-52-46(67-52)31-22-13-10-16-25-34-55(58)61-40-42(63-57(60)36-27-18-12-15-24-33-48-54(69-48)39-51-45(66-51)30-21-9-6-3)41-62-56(59)35-26-17-11-14-23-32-47-53(68-47)38-50-44(65-50)29-20-8-5-2/h42-54H,4-41H2,1-3H3
LEDER O=C(OCC(OC(CCCCCCCC1OC1CC2OC2CCCCC)=O)COC(CCCCCCCC3OC3CC4OC4CCCCC)=O)CCCCCCCC5OC5CC6OC6CCCCC
IUPAC-namn 2,3-Bis[8-[3-[(3-pentyloxiran-2-yl)methyl]oxiran-2-yl]octanoyloxy]propyl 8-[3-[(3-pentyloxiran-2-yl)methyl]oxiran-2-yl]octanoate
Molekylvikt (g/mol) 975.4
Formel vikt 974.71