Läs mer
Tocris Bioscience™ SP 100030
NF-κB and AP-1 dual inhibitor
Brand: Tocris Bioscience™ 5309/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SP 100030 is a potent dual inhibitor of NF-κB and AP-1 transcriptional activity (IC50 = 50 nM). Blocks production of IL-2, IL-8 and TNF-α from Jurkat T cells. Inhibits cytokine production selectively in T cells; exhibits minimal inhibition of cytokine production in other cell types. Decreases arthritic severity in a mouse model of collagen-induced arthritis. Also ameliorates muscle wasting in a rat model of cancer cachexia.Specifikationer
| N-[3,5-Bis(trifluoromethyl)phenyl]-2-chloro-4-(trifluoromethyl)-5-pyrimidinecarboxamide | |
| 154563-54-9 | |
| 50 mg |