missing translation for 'onlineSavingsMsg'
Learn More

SP1 Mouse anti-Human, Clone: 4G3H3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66508-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878445

  • 4721.67 SEK / 150µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SP1 is a transcription factor that can activate or repress transcription in response to physiological and pathological stimuli. Binds with high affinity to GC-rich motifs and regulates the expression of a large number of genes involved in a variety of processes such as cell growth, apoptosis, differentiation and immune responses. Highly regulated by post-translational modifications (phosphorylations, sumoylation, proteolytic cleavage, glycosylation and acetylation). Binds also the PDGFR-alpha G-box promoter. May have a role in modulating the cellular response to DNA damage. Implicated in chromatin remodeling. Plays a role in the recruitment of SMARCA4/BRG1 on the c-FOS promoter. Plays an essential role in the regulation of FE65 gene expression.
Specifications

Specifications

SP1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08047
SP1
SP1 Fusion Protein Ag16720
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
4G3H3
Unconjugated
SP1
SP1, Sp1 transcription factor, Transcription factor Sp1, TSFP1
Mouse
Protein G
RUO
6667
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers