missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP1 Mouse anti-Human, Clone: 4G3H3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66508-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
SP1 is a transcription factor that can activate or repress transcription in response to physiological and pathological stimuli. Binds with high affinity to GC-rich motifs and regulates the expression of a large number of genes involved in a variety of processes such as cell growth, apoptosis, differentiation and immune responses. Highly regulated by post-translational modifications (phosphorylations, sumoylation, proteolytic cleavage, glycosylation and acetylation). Binds also the PDGFR-alpha G-box promoter. May have a role in modulating the cellular response to DNA damage. Implicated in chromatin remodeling. Plays a role in the recruitment of SMARCA4/BRG1 on the c-FOS promoter. Plays an essential role in the regulation of FE65 gene expression.Specifications
SP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08047 | |
SP1 | |
SP1 Fusion Protein Ag16720 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
4G3H3 | |
Unconjugated | |
SP1 | |
SP1, Sp1 transcription factor, Transcription factor Sp1, TSFP1 | |
Mouse | |
Protein G | |
RUO | |
6667 | |
-20°C | |
Liquid |