Learn More
SP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21962-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
SP1 is a transcription factor that can activate or repress transcription in response to physiological and pathological stimuli. Binds with high affinity to GC-rich motifs and regulates the expression of a large number of genes involved in a variety of processes such as cell growth, apoptosis, differentiation and immune responses. Highly regulated by post-translational modifications (phosphorylations, sumoylation, proteolytic cleavage, glycosylation and acetylation). Binds also the PDGFR-alpha G-box promoter. May have a role in modulating the cellular response to DNA damage. Implicated in chromatin remodeling. Plays a role in the recruitment of SMARCA4/BRG1 on the c-FOS promoter. Plays an essential role in the regulation of FE65 gene expression.Specifications
SP1 | |
Polyclonal | |
Unconjugated | |
SP1 | |
SP1, Sp1 transcription factor, Transcription factor Sp1, TSFP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20683, 24790, 6667 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O89090, P08047, Q01714 | |
SP1 | |
SP1 Fusion Protein Ag16720 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |