Läs mer
SP187, MedChemExpress
Produktkod. 16482408
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
20 mg
Förpackningsstorlek:
1mg
20mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16482408 5 mg -
16470209 10 mg -
16450726 20 mg 20mg
16413199 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16482408 Leverantör Medchem Express Leverantörsnummer HYU00160
MedChemExpress SP187 is a host-targeted iminosugar with activity against filovirus infections in vitro and in vivo. SP187 is active against influenza and dengue in vivo.
SP187 is a host-targeted iminosugar with activity against filovirus infections in vitro and in vivo. SP187 is active against influenza and dengue in vivo.
Specifikationer
Kemiskt namn eller material SP187
Fysisk form Solid
CAS 615253-61-7
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C16H33NO5
För användning med (applikation) COVID-19-anti-virus
Synonym MON-​DNJ UV4
Löslighetsinformation DMSO : 100 mg/mL (313.05 mM; Need ultrasonic)
LEDER OC[C@H]1N(C[C@H](O)[C@@H](O)[C@@H]1O)CCCCCCCCCOC
Molekylvikt (g/mol) 319.44
Formel vikt 319.44
Procent renhet 98.0%