Learn More
Alfa Aesar™ SP6 RNA Polymerase in Potassium Phosphate Buffer Kit
Transcribes RNA from DNA sequences
Brand:  Alfa Aesar™ J65573.O@
Additional Details : CAS Number : 9014-24-8 Weight : 5.00000kg
 View more versions of this product
Product Code. 15496969
  • 16708.41 SEK / 5kg
Estimated Shipment date
from Supplier 18-02-2019
Add to basket
Specifications
Specifications
Colorless
Liquid
5 Kilounits
9014-24-8
Transcribes RNA from DNA sequences
RNA Polymerase
−30° to −10°C
RNA Polymerase from Salmonella enterica serotype typhimurium, SP6-infected