Learn More
SPACA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554852
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TNNGIFQINS RRWCSNLTPN VPNVCRMYCS DLLNPNLKDT VICAMKITQE PQGLGYWEAW RHHCQGKDLT EWVDGCDF Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.
SPACA3 Polyclonal antibody specifically detects SPACA3 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SPACA3 | |
Polyclonal | |
Unconjugated | |
SPACA3 | |
1700025M08Rik; acrosomal sperm protein; ALLP17; Cancer/testis antigen 54; CT54; LYC3; lysozyme C; lysozyme-like acrosomal sperm-specific secretory protein ALLP-17; Lysozyme-like protein 3; lysozyme-like sperm-specific secretory protein ALLP17; LYZC; Lyzl3; mSLLP1; SLLP1; Spaca3; sperm acrosome associated 3; sperm acrosome membrane-associated protein 3; Sperm acrosome membrane-associated protein 3, membrane form; Sperm acrosome membrane-associated protein 3, processed form; sperm c-type lysozyme-like protein; sperm lysozyme like protein 1; sperm lysozyme-like protein 1; sperm lysozyme-like protein 1 precusor; sperm lyzozyme-like acrosomal protein 1; Sperm protein reactive with antisperm antibodies; Sperm protein reactive with ASA; SPRASA; UNQ424/PRO862 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124912 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXA5 | |
SPACA3 | |
Recombinant protein corresponding to Human SPACA3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only