missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550556
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total SPAG6 protein.
SPAG6 Polyclonal antibody specifically detects SPAG6 in Human, Mouse samples. It is validated for Western Blot.Specifications
SPAG6 | |
Polyclonal | |
Unconjugated | |
SPAG6 | |
axoneme central apparatus protein; CT141; PF16; protein PF16 homolog; Repro-SA-1; RGD1310892; SPAG6; Spag6l; sperm associated antigen 6; sperm associated antigen 6 like; sperm flagellar protein; Sperm-associated antigen 6; testicular tissue protein Li 176 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
381350, 9576 | |
-20°C | |
Liquid |
Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O75602, Q9JLI7 | |
SPAG6 | |
Synthetic peptide corresponding to residues near the N terminal of human sperm associated antigen 6 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |