Learn More
Abnova™ SPARCL1 Recombinant Protein
Human SPARCL1 full-length ORF recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH33721 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 8404 |
Molecular Weight (g/mol) | 96.91 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16172422
|
Abnova™
H00008404-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182422
|
Abnova™
H00008404-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Theoretical MW (kDa): 96.91
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: PTNARLLSDHSKPTAETVAPDNTAIPSLRAEDEENEKETAVSTEDDSHHKAEKSSVLKSKEESHEQSAEQGKSSSQELGLKDQEDSDGDLSVNLEYAPTEGTLDIKEDMSEPQEKKLSENTDFLAPGVSSFTDSNQQESITKREENQEQPRNYSHHQLNRSSKHSQGLRDQGNQEQDPNISNGEEEEEKEPGEVGTHNDNQERKTELPREHANSKQEEDNTQSDDILEESDQPTQVSKMQEDEFDQGNQEQEDNSNAEMEEENASNVNKHIQETEWQSQEGKTGLEAISNHKETEEKTVSEALLMEPTDDGNTTPRNHGVDDDGDDDGDDGGTDGPRHSASDDYFIPSQAFLEAERAQSIAYHLKIEEQREKVHENENIGTTEPGEHQEAKKAENSSNEEETSSEGNMRVHAVDSCMSFQCKRGHICKADQQGKPHCVCQDPVTCPPTKPLDQVCGTDNQTYASSCHLFATKCRLEGTKKGHQLQLDYFGACKSIPTCTDFEVIQFPLRMRDWLKNILMQLYEANSEHAGYLNEKQRNKVKKIYLDEKRLLAGDHPIDLLLRDFKKNYHMYVYPVHWQFSELDQHPMDRVLTHSELAPLRASLVPMEHCITRFFEECDPNKDKHITLKEWGHCFGIKEEDIDENLLF
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH33721 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
96.91 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SPARCL1 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
8404 | |
SPARCL1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PTNARLLSDHSKPTAETVAPDNTAIPSLRAEDEENEKETAVSTEDDSHHKAEKSSVLKSKEESHEQSAEQGKSSSQELGLKDQEDSDGDLSVNLEYAPTEGTLDIKEDMSEPQEKKLSENTDFLAPGVSSFTDSNQQESITKREENQEQPRNYSHHQLNRSSKHSQGLRDQGNQEQDPNISNGEEEEEKEPGEVGTHNDNQERKTELPREHANSKQEEDNTQSDDILEESDQPTQVSKMQEDEFDQGNQEQEDNSNAEMEEENASNVNKHIQETEWQSQEGKTGLEAISNHKETEEKTVSEALLMEPTDDGNTTPRNHGVDDDGDDDGDDGGTDGPRHSASDDYFIPSQAFLEAERAQSIAYHLKIEEQREKVHENENIGTTEPGEHQEAKKAENSSNEEETSSEGNMRVHAVDSCMSFQCKRGHICKADQQGKPHCVCQDPVTCPPTKPLDQVCGTDNQTYASSCHLFATKCRLEGTKKGHQLQLDYFGACKSIPTCTDFEVIQFPLRMRDWLKNILMQLYEANSEHAGYLNEKQRNKVKKIYLDEKRLLAGDHPIDLLLRDFKKNYHMYVYPVHWQFSELDQHPMDRVLTHSELAPLRASLVPMEHCITRFFEECDPNKDKHITLKEWGHCFGIKEEDIDENLLF | |
PIG33/SC1 | |
SPARCL1 | |
Wheat Germ (in vitro) | |
GST |