Learn More
Duran™ Spare Glass Pipettes
Click to view available options
Capacity (Metric):
50 mL
100 mL
Specifications
Product Type | Spare Pipette |
Capacity (Metric) | 50 mL |
Color | Clear |
Material | Glass |
Quantity | 10Pack |