missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17357-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a testis-specific protein that belongs to the tetratricopeptide repeat-like superfamily. The encoded protein localizes to the Golgi apparatus and may play a role in spermatogenesis.Specifications
SPATA16 | |
Polyclonal | |
Unconjugated | |
SPATA16 | |
NYD SP12, SPATA16, spermatogenesis associated 16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294932, 70862, 83893 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C636, Q9BXB7 | |
Spata16 | |
SPATA16 Fusion Protein Ag10637 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |