missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA19 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16656-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May have a role in spermiogenesis.Specifications
SPATA19 | |
Polyclonal | |
Unconjugated | |
SPATA19 | |
SPAS1, SPATA19, Spergen 1, spergen1, spermatogenesis associated 19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
219938 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7Z5L4 | |
SPATA19 | |
SPATA19 Fusion Protein Ag10108 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |