missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPDYA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17836-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Speedy A, also known as SPDYA, SPDY1, Ringo3 or SPY1, is a 313 amino acid protein that localizes to the nucleus and belongs to the speedy/ringo family. Expressed at high levels in testis and at lower levels in brain, kidney, heart, bone marrow, colon, lung, liver spleen and placenta, Speedy A functions to regulate the G1/S phase transition of the cell cycle, specifically by binding to and activating Cdc2 p34, Cdk2 and p27. Additionally, Speedy A mediates cell survival during DNA damage, suggesting that Speedy A plays a role in proper cell cycle progression throughout the life of the cell. Multiple isoforms of Speedy A exist due to alternative splicing events. The gene encoding Speedy A maps to human chromosome 2, which encodes over 1,400 genes and comprises nearly 8% of the human genome.Specifications
SPDYA | |
Polyclonal | |
Unconjugated | |
SPDYA | |
hSpy/Ringo A, RINGO A, Ringo3, SPDY1, SPDYA, Speedy 1, Speedy protein A, SPY1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192209, 245711, 70891 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5IBH7, Q5MJ70, Q8R496 | |
Spdya | |
SPDYA Fusion Protein Ag12041 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |