missing translation for 'onlineSavingsMsg'
Learn More

SPE tube ODS reversed phase polypropylene 1g, 6mL Bakerbond

Brand:  JT Baker 7020-27

Additional Details : CAS Number : 71889-02-6 Weight : 3.00000kg

Product Code. 15578384

  • 20530.00 SEK / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket