Läs mer
3M™ Speaking Diaphragm Kit, 7895
Handla allt 3M Produkter
:
For Use With: 3M Full Facepieces 7000 Series
Beskrivning
Replace old or damaged 3M™ Powerflow™ Full Facepiece 7900 speech diaphragms with 3M™ Speaking Diaphragm Kit, 7895. Our approved 3M replacement part can extend the service life of your facepiece. This kit includes speaking diaphragm, gasket, retainer and removal tool.
Specifikationer
Specifikationer
| Certifikat och efterlevnad | NIOSH Approved |
| Produkttyp | Replacement Speaking Diaphragm Kit |
| För användning med (utrustning) | 3M™ Full Facepieces 7000 Series |