Läs mer
3M™ Specialty Particulate Respirator 9906
Produktkod. 15749837 Handla allt 3M Produkter
Change view
Klicka för att se tillgängliga alternativ
Color: White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod.
15749837 Color: White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 15749837 Leverantör 3M™ Leverantörsnummer 7000088785
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
3M™ Specialty Particulate Respirator 9906 provides lightweight FFP1 protection against dusts, mists, and hydrogen fluoride.
  • Provides FFP1 (NPF 4) protection against dusts and mists
  • Activated carbon layer helps guard against nuisance-level hydrogen fluoride
  • Convex shape, nose clip and twin strapping helps provide a comfortable fit across a broad range of face shapes
  • Lightweight, cup-shaped design is sturdy and collapse resistant
  • Braided, color-coded headbands help you identify the protection level offered: yellow for FFP1
  • Designed to work well with 3M™ Eyewear and Hearing Protection products
  • Good fit over a wide range of face sizes provided by convex shape, nose clip and twin-strap design
TRUSTED_SUSTAINABILITY
Specifikationer
Certifikat och efterlevnad CE Approved, EN 149: 2001 + A1: 2009 FFP1 NR D
Färg White
Filtertyp FFP1 Protection Level
Produkttyp Specialty Particulate Respirator
Med utandningsventil No
Motståndskraftig mot <TLV for Hydrogen Fluoride, Up to 4 x TLV for Particulates