Learn More
3M™ Specialty Particulate Respirator 9914SP
3M™ Specialty Particulate Respirator 9914SP provides lightweight FFP1 protection against dusts, mists, and organic vapors.
Brand: 3M™ 7100146835
Additional Details : Weight : 2.24000kg
Description
- Provides FFP1 (NPF 12) protection against dusts, mists, and organic vapors
- Activated carbon layer provides relief against nuisance-level odors
- 3M™ Cool Flow™ Valve helps reduce heat build-up
- Convex shape, nose clip and twin strapping helps provide a comfortable fit across a broad range of face shapes
- Lightweight, cup-shaped design is sturdy and collapse resistant
- Braided, color-coded headbands help you identify the protection level offered: yellow for FFP1
- Small pack size of 5. Larger pack size available
- Good fit over a wide range of face sizes provided by convex shape, nose clip and twin-strap design
Specifications
CE Approved, EN 149: 2001 + A1: 2009 FFP1 NR D | |
FFP1 Protection Level | |
5/Pk. |
Gray | |
Yes | |
<TLV for Organic Vapor , Up to 4 x TLV for Particulates |