Läs mer
Speciociliatine, TRC
Produktkod. 30011917
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011917 1 mg
30006804 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011917 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6801001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14382-79-7
Molekylformel C23 H30 N2 O4
Molekylvikt (g/mol) 398.5
Synonym 17,18-Seco-3β,20α-yohimban-16-carboxylic acid, 16,17-didehydro-9,17-dimethoxy-, methyl ester, (E)- (8CI), Corynan-16-carboxylic acid, 16,17-didehydro-9,17-dimethoxy-, methyl ester, (3β,16E,20β)-, Speciociliatin, Speciociliatine, Indolo[2,3-a]quinolizine-2-acetic acid, 3-ethyl-1,2,3,4,6,7,12,12b-octahydro-8-methoxy-α-(methoxymethylene)-, methyl ester, (αE,2S,3S,12bR)-
IUPAC-namn methyl (E)-2-[(2S,3S,12bR)-3-ethyl-8-methoxy-1,2,3,4,6,7,12,12b-octahydroindolo[2,3-a]quinolizin-2-yl]-3-methoxyprop-2-enoate
LEDER CC[C@@H]1CN2CCc3c([nH]c4cccc(OC)c34)[C@H]2C[C@@H]1\C(=C/OC)\C(=O)OC
Specifikationer
Kemiskt namn eller material Speciociliatine
Kvantitet 1 mg
InChI formel InChI=1S/C23H30N2O4/c1-5-14-12-25-10-9-15-21-18(7-6-8-20(21)28-3)24-22(15)19(25)11-16(14)17(13-27-2)23(26)29-4/h6-8,13-14,16,19,24H,5,9-12H2,1-4H3/b17-13+/t14-,16+,19-/m1/s1
Formel vikt 398.2206