Learn More
Spectinomycin Sulfate Tetrahydrate, TRC
Product Code. 30137098
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137098 10 g 10g
30135451 1 g 1g
30005442 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137098 Supplier Toronto Research Chemicals Supplier No. TRCS68025310G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64058-48-6
Molecular Formula C14 H24 N2 O7 . H2 O . H2 O4 S
Molecular Weight (g/mol) 448.44
Synonym 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one, decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-, (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-, sulfate (1:1), 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one, decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-, (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-, sulfate (1:1) (salt) (9CI), 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one, decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-, [2R-(2α,4aβ,5aβ,6β,7β,8β,9α,9aα,10aβ)]-, sulfate (1:1) (salt), 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one, decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-, sulfate (1:1) (salt) (8CI), Actinospectacin sulfate, Spectinomycin sulfate
SMILES O.O.O.O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O.OS(=O)(=O)O
Specifications
Quantity 10 g
InChI Formula InChI=1S/C14H24N2O7.H2O4S.4H2O/c1-5-4-6(17)14(20)13(21-5)22-12-10(19)7(15-2)9(18)8(16-3)11(12)23-14;1-5(2,3)4;;;;/h5,7-13,15-16,18-20H,4H2,1-3H3;(H2,1,2,3,4);4*1H2/t5-,7-,8+,9+,10+,11-,12-,13+,14+;;;;;/m1...../s1
Formula Weight 448.4433
Chemical Name or Material Spectinomycin sulfate tetrahydrate