Learn More
Spectrum Labs™ Spectra/Chrom™ Aqueous Series Columns
3420.00 SEK - 7655.00 SEK
Specifications
Type | Aqueous Column |
---|---|
For Use With (Equipment) | For use with standard buffers |
Quantity | 1 Each |
Material | Polypropylene |
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
11979328
|
Spectrum Labs™
124022 |
25 mm | 300 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11909338
|
Spectrum Labs™
124036 |
50 mm | 300 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11919338
|
Spectrum Labs™
124038 |
50 mm | 600 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Aqueous Column | |
For use with standard buffers | |
1 Each | |
Polypropylene |