Läs mer
Molecular Devices SpectraMax™ ABS Single‐Mode Microplate Reader with SoftMax Pro Software
Handla allt Molecular Devices ProdukterBeskrivning
- Designed for absorbance detection workflows in microplate-based assays
- Supports UV-Vis absorbance measurements for life science applications
- Enables flexible assay setup with configurable measurement parameters
- Supports microplate-based assay workflows across multiple applications
- Integrated with SoftMax Pro Software for data acquisition and analysis
- Supports routine absorbance-based assays with consistent and reproducible performance
- Designed for use in laboratory environments requiring reliable microplate reading
Specifikationer
Specifikationer
| Detektortyp | Photodiode |
| Gränssnitt | PC Control, Internal Software |
| Ljuskälla | Xenon Flash Lamp |
| Mätteknik | Absorbance (Top) |
| Typ av tallrik | 96 and 384-well Plates |
| Läsalternativ | Endpoint |
| Skakning | None |
| Val av våglängd | Monochromator |