Läs mer
Molecular Devices SpectraMax™ i3x Multi‐Mode Microplate Detection Platform with SoftMax Pro Software
Handla allt Molecular Devices ProdukterBeskrivning
- Designed for multi-mode detection workflows in microplate-based assays
- Supports a range of detection capabilities for diverse life science applications
- Enables flexible assay setup with configurable detection parameters
- Supports biochemical and cell-based assay workflows
- Provides expandable functionality through modular detection options
- Integrated with SoftMax Pro Software for data acquisition and analysis
- Supports scalable workflows for research and laboratory environments
Specifikationer
Specifikationer
| Inkubatortemperatur | 4-Zone Incubation to 45°C |
| Mäthastighet | 11 sec. (96 wells), 22 sec. (384 wells) |
| Skakning | Linear, Orbital |
| Typ av tallrik | 6 to 384-well Plates |
| Detektortyp | PMT |
| Programvarutyp | SoftMax Pro |
| Ljuskälla | Xenon Flash |
| Läsalternativ | Endpoint, Kinetic, Spectral Scanning |