Learn More
3M™ Speedglas™ Outer Protection Plate 9000
Product Code. 15799627 Shop All 3M Products
Change view
Click to view available options
Style:
Heat Resistant
Scratch Resistant
Standard
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Style Unit Size
15799627 Scratch Resistant Pack of 10
15729637 Heat Resistant Pack of 10
15780048 Standard Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15799627 Supplier 3M™ Supplier No. 7000000183
  • ONLINE EXCLUSIVE PRICE
    822.00 SEK
    668.54 SEK / Paket med 10
    Save 153.46 SEK
    18% Av
Beräknad leverans: 25-01-2027
Add to basket
Add to basket
Denne vare kan ikke returneres. Se returpolicy
3M™ Speedglas™ Outer Protection Plate 9000 for use with 3M™ Speedglas™ Welding Helmet Series 9000.
  • Replacement spare part to the original protection plate if any pits or scratches occur
  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Regularly clean with a soft cloth and replace if any pits or scratches occur
TRUSTED_SUSTAINABILITY
Tekniske data
Certifications/Compliance EN 166‎, EN166:BT
For Use With (Equipment) Speedglas Welding Helmet Series 9000
Lens Tint Clear
Product Type Helmet Outside Protection Plate
Style Scratch Resistant
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.