missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPESP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28010-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The encoded protein is a human alloantigen involved in sperm-egg binding and fusion.Specifications
SPESP1 | |
Polyclonal | |
Unconjugated | |
SPESP1 | |
Equatorial segment protein, ESP, SP ESP, SPESP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246777, 501010, 66712 | |
-20°C | |
Liquid |
Western Blot | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A0JPP4, Q6UW49, Q9D5A0 | |
Spesp1 | |
SPESP1 Fusion Protein Ag27764 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |