missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPESP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SPESP1 |
---|---|
Concentration | 0.6 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825955
|
Proteintech
28010-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815955
|
Proteintech
28010-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The encoded protein is a human alloantigen involved in sperm-egg binding and fusion.Specifications
SPESP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A0JPP4, Q6UW49, Q9D5A0 | |
246777, 501010, 66712 | |
SPESP1 Fusion Protein Ag27764 | |
Primary | |
-20°C | |
SPESP1 |
0.6 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Equatorial segment protein, ESP, SP ESP, SPESP1 | |
Spesp1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |