Learn More
SPEX CertiPrep™ Method AOAC 2007.01 SPEXQuE™ QuEChERS Kit
Sensible Solutions for Pesticide Extraction
1890.00 SEK
Product Code | Brand | Analyte or Component Names | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Analyte or Component Names | Quantity | Price | Quantity & Availability | ||||
15255866
|
SPEX Certiprep
AOAC-PSA-15ML |
400mg PSA, 1200mg MgSO4 | 15 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.
- US (AOAC Method 2007.01)