Learn More
SPEX CertiPrep™ Method AOAC 2007.01 SPEXQuE™ QuEChERS Kit
Sensible Solutions for Pesticide Extraction
1890.00 SEK
Products 1
Product Code Brand Analyte or Component Names Quantity Price Quantity & Availability  
Product Code Brand Analyte or Component Names Quantity Price Quantity & Availability  
15255866
View Documents Promotion Details
SPEX Certiprep
AOAC-PSA-15ML
400mg PSA, 1200mg MgSO4 15 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.
  • US (AOAC Method 2007.01)