Learn More
SPEXQuE™ AOAC QC Spike Mix, SPEX CertiPrep™
Sensible solutions for pesticide extraction
Brand: SPEX Certiprep QUECR-AOAC-QC
Additional Details : Weight : 0.00100kg Transport : UN number : 1648 Chem class : 3 Pack group : II
Description
- Solvent: Acetonitrile
- Volume: 5mL
Specifications
40 µg/mL | |
5 mL | |
AOAC Internal Standard Mix |
Acetonitrile | |
Pesticide Extraction |